Anti-PITPNB Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5858
BWRLBS5858EA 262 EUR
BWRLBS5858
Anti-PITPNB Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 226-271 of Human PITPNB.

PITPNB pAb detects endogenous levels of PITPNB protein.

Type: Primary
Antigen: PITPNB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR