Anti-PRKX Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5865
BWRLBS5865EA 262 EUR
BWRLBS5865
Anti-PRKX Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 308-353 of Human PRKX.

PRKX pAb detects endogenous levels of PRKX protein.

Type: Primary
Antigen: PRKX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR