Anti-RANBP6 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5882
BWRLBS5882EA 262 EUR
BWRLBS5882
Anti-RANBP6 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 841-886 of Human RANBP6.

RANBP6 pAb detects endogenous levels of RANBP6 protein.

Type: Primary
Antigen: RANBP6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR