Anti-ARHGEF1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5901
BWRLBS5901EA 262 EUR
BWRLBS5901
Anti-ARHGEF1 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 206-251 of Human ARHGEF1.

ARHGEF1 pAb detects endogenous levels of ARHGEF1 protein.

Type: Primary
Antigen: ARHGEF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR