Anti-RPL17 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5903
BWRLBS5903EA 262 EUR
BWRLBS5903
Anti-RPL17 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 139-184 of Human RPL17.

RPL17 pAb detects endogenous levels of RPL17 protein.

Type: Primary
Antigen: RPL17
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR