Anti-PPP1R19 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5913
BWRLBS5913EA 262 EUR
BWRLBS5913
Anti-PPP1R19 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 181-226 of Human PPP1R19.

PPP1R19 pAb detects endogenous levels of PPP1R19 protein.

Type: Primary
Antigen: PPP1R19
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR