Anti-RORgamma Rabbit Polyclonal Antibody
Lieferant: Bioworld Technology
BS5915
BWRLBS5915EA 262 EUR
BWRLBS5915
Anti-RORgamma Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 114-159 of Human RORgamma.
ROR? pAb detects endogenous levels of ROR? protein.
Type: Primary
Antigen: RORgamma
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat