Anti-RTCD1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5918
BWRLBS5918EA 262 EUR
BWRLBS5918
Anti-RTCD1 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 321-366 of Human RTCD1.

RTCD1 pAb detects endogenous levels of RTCD1 protein.

Type: Primary
Antigen: RTCD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR