Anti-SLC10A7 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5931
BWRLBS5931EA 262 EUR
BWRLBS5931
Anti-SLC10A7 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 212-257 of Human SLC10A7.

SLC10A7 pAb detects endogenous levels of SLC10A7 protein.

Type: Primary
Antigen: SLC10A7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR