Anti-SSBP2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5946
BWRLBS5946EA 262 EUR
BWRLBS5946
Anti-SSBP2 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 2-47 of Human SSBP2.

SSBP2 pAb detects endogenous levels of SSBP2 protein.

Type: Primary
Antigen: SSBP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR