Anti-STMN2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5948
BWRLBS5948EA 262 EUR
BWRLBS5948
Anti-STMN2 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 134-179 of Human STMN2.

STMN2 pAb detects endogenous levels of STMN2 protein.

Type: Primary
Antigen: STMN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR