Anti-SUHW1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5951
BWRLBS5951EA 262 EUR
BWRLBS5951
Anti-SUHW1 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 491-536 of Human SUHW1.

SUHW1 pAb detects endogenous levels of SUHW1 protein.

Type: Primary
Antigen: SUHW1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR