Anti-UBASH3A Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5968
BWRLBS5968EA 262 EUR
BWRLBS5968
Anti-UBASH3A Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 356-401 of Human UBASH3A.

UBASH3A pAb detects endogenous levels of UBASH3A protein.

Type: Primary
Antigen: UBASH3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR