Anti-VHL Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS60046
BWRLBS60046EA 262 EUR
BWRLBS60046
Anti-VHL Rabbit Polyclonal Antibody
Antikörper
A synthetic peptide corresponding to residues in Human VHL.

VHL pAb detects endogenous levels of VHL protein.

Type: Primary
Antigen: VHL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR