Anti-ID1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS60047
BWRLBS60047EA 262 EUR
BWRLBS60047
Anti-ID1 Rabbit Polyclonal Antibody
Antikörper
A synthetic peptide corresponding to residues in Human ID1.

ID1 pAb detects endogenous levels of ID1 protein.

Type: Primary
Antigen: ID1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR