Anti-SNRPD3 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS60048
BWRLBS60048EA 262 EUR
BWRLBS60048
Anti-SNRPD3 Rabbit Polyclonal Antibody
Antikörper
A synthetic peptide corresponding to residues in Human SNRPD3.

SNRPD3 pAb detects endogenous levels of SNRPD3 protein.

Type: Primary
Antigen: SNRPD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR