Anti-POLR2H Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS60148
BWRLBS60148EA 262 EUR
BWRLBS60148
Anti-POLR2H Rabbit Polyclonal Antibody
Antikörper
A synthetic peptide corresponding to residues in Human POLR2H.

POLR2H pAb detects endogenous levels of POLR2H protein.

Type: Primary
Antigen: POLR2H
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR