Anti-Glut 2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS60254
BWRLBS60254EA 262 EUR
BWRLBS60254
Anti-Glut 2 Rabbit Polyclonal Antibody
Antikörper
A synthetic peptide corresponding to residues in Human Glut 2.

Glut 2 pAb detects endogenous levels of Glut 2 protein.

Type: Primary
Antigen: Glut 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR