Anti-INDO Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS60456
BWRLBS60456EA 262 EUR
BWRLBS60456
Anti-INDO Rabbit Polyclonal Antibody
Antikörper
A synthetic peptide corresponding to residues in Human INDO.

INDO pAb detects endogenous levels of INDO protein.

Type: Primary
Antigen: INDO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR