Anti-Rho C Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6113
BWRLBS6113EA 341 EUR
BWRLBS6113
Anti-Rho C Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human Rho C.

RHOC pAb detects endogenous levels of RHOC protein.

Type: Primary
Antigen: Rho C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR