Anti-GDF-15 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6152
BWRLBS6152EA 341 EUR
BWRLBS6152
Anti-GDF-15 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human GDF-15.

GDF15 pAb detects endogenous levels of GDF15 protein.

Type: Primary
Antigen: GDF-15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR