Anti-EMD Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6162
BWRLBS6162EA 341 EUR
BWRLBS6162
Anti-EMD Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human EMD.

EMD pAb detects endogenous levels of EMD protein.

Type: Primary
Antigen: EMD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR