Anti-PEBP1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6176
BWRLBS6176EA 341 EUR
BWRLBS6176
Anti-PEBP1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human PEBP1.

PEBP1 pAb detects endogenous levels of PEBP1 protein.

Type: Primary
Antigen: PEBP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR