Anti-SCRN2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6246
BWRLBS6246EA 341 EUR
BWRLBS6246
Anti-SCRN2 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human SCRN2.

SCRN2 pAb detects endogenous levels of SCRN2 protein.

Type: Primary
Antigen: SCRN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol SCRN2
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Polyklonal
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Isotyp
Western blot Yes
ImmunoChemie Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR