Anti-p-Raf-B Rabbit Polyclonal Antibody [clone: S445]

Lieferant: Bioworld Technology
BS6338
BWRLBS6338EA 379 EUR
BWRLBS6338
Anti-p-Raf-B Rabbit Polyclonal Antibody [clone: S445]
Antikörper
Synthetic phosphopeptide derived from human Raf-B around the phosphorylation site of Serine 445.

P-Raf-B (S445) pAb detects endogenous levels of Raf-B protein when phosphorylated at Ser445.

Type: Primary
Antigen: p-Raf-B
Clonality: Polyclonal
Clone: S445
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR