Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: T161 ]

Lieferant: Bioworld Technology
BS6340
BWRLBS6340EA 379 EUR
BWRLBS6340
Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: T161 ]
Antikörper
Synthetic phosphopeptide derived from human Cdk1/Cdc2 around the phosphorylation site of Serine 795.

p-Cdk1/Cdc2 (T161 ) pAb detects endogenous levels of Cdk1/Cdc2 protein only when phosphorylated at Thr161.

Type: Primary
Antigen: p-Cdk1/Cdc2
Clonality: Polyclonal
Clone: T161
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR