Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: Y15]

Lieferant: Bioworld Technology
BS6342
BWRLBS6342EA 379 EUR
BWRLBS6342
Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: Y15]
Antikörper
Synthetic phosphopeptide derived from human Cdk1/Cdc2 around the phosphorylation site of Serine 795.

p-Cdk1/Cdc2 (T15) pAb detects endogenous levels of Cdk1/Cdc2 protein only when phosphorylated at Thr15.

Type: Primary
Antigen: p-Cdk1/Cdc2
Clonality: Polyclonal
Clone: Y15
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR