Anti-ASK1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6445
BWRLBS6445EA 341 EUR
BWRLBS6445
Anti-ASK1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human ASK1.

ASK1 pAb detects endogenous levels of ASK1 protein.

Type: Primary
Antigen: ASK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR