Anti-HSP90alpha Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6461
BWRLBS6461EA 341 EUR
BWRLBS6461
Anti-HSP90alpha Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human HSP90alpha.

HSP90? pAb detects endogenous levels of HSP90? protein.

Type: Primary
Antigen: HSP90alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR