Anti-CDK4 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6462
BWRLBS6462EA 341 EUR
BWRLBS6462
Anti-CDK4 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human CDK4.

CDK4 pAb detects endogenous levels of CDK4 protein.

Type: Primary
Antigen: CDK4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR