Anti-MEK2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6472
BWRLBS6472EA 341 EUR
BWRLBS6472
Anti-MEK2 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human MEK2.

MEK2 pAb detects endogenous levels of MEK2 protein.

Type: Primary
Antigen: MEK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR