Anti-ADORA2A Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6513
BWRLBS6513EA 341 EUR
BWRLBS6513
Anti-ADORA2A Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human ADORA2A.

ADORA2A pAb detects endogenous levels of ADORA2A protein.

Type: Primary
Antigen: ADORA2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR