Anti-p21 Rabbit Polyclonal Antibody [clone: FL]

Lieferant: Bioworld Technology
BS6561
BWRLBS6561EA 341 EUR
BWRLBS6561
Anti-p21 Rabbit Polyclonal Antibody [clone: FL]
Antikörper
Recombinant full length Human P21.

p21 (FL) pAb detects endogenous levels of p21 protein.

Type: Primary
Antigen: p21
Clonality: Polyclonal
Clone: FL
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR