Anti-mAChR M2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6566
BWRLBS6566EA 341 EUR
BWRLBS6566
Anti-mAChR M2 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human mAChR M2.

CHRM2 pAb detects endogenous levels of CHRM2 protein.

Type: Primary
Antigen: mAChR M2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR