Anti-PPP4C Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6703
BWRLBS6703EA 341 EUR
BWRLBS6703
Anti-PPP4C Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human PPP4C.

PPP4C pAb detects endogenous levels of PPP4C protein.

Type: Primary
Antigen: PPP4C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR