Anti-Renin Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6722
BWRLBS6722EA 341 EUR
BWRLBS6722
Anti-Renin Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human Renin.

Renin pAb detects endogenous levels of Renin protein.

Type: Primary
Antigen: Renin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR