Anti-TAP2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6846
BWRLBS6846EA 341 EUR
BWRLBS6846
Anti-TAP2 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human TAP2.

  • TAP2 pAb detects endogenous levels of TAP2 protein.

Type: Primary
Antigen: TAP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR