Anti-DNA pol beta Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6855
BWRLBS6855EA 341 EUR
BWRLBS6855
Anti-DNA pol beta Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human DNA pol beta.

DNA pol ? pAb detects endogenous levels of DNA pol ? protein.

Type: Primary
Antigen: DNA pol beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR