Anti-MMP9 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6893
BWRLBS6893EA 341 EUR
BWRLBS6893
Anti-MMP9 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human MMP9.

MMP9 pAb detects endogenous levels of MMP9 protein.

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR