Anti-CA1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6959
BWRLBS6959EA 341 EUR
BWRLBS6959
Anti-CA1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human CA1.

CA1 pAb detects endogenous levels of CA1 protein.

Type: Primary
Antigen: CA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR