Anti-BCHE Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS6965
BWRLBS6965EA 341 EUR
BWRLBS6965
Anti-BCHE Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human BCHE.

BCHE pAb detects endogenous levels of Cholinesterase protein.

Type: Primary
Antigen: BCHE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR