Anti-eIF4A1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7106
BWRLBS7106EA 341 EUR
BWRLBS7106
Anti-eIF4A1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human eIF4A1.

eIF4A1 pAb detects endogenous levels of eIF4A1 protein.

Type: Primary
Antigen: eIF4A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR