Anti-Ephrin-A1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7110
BWRLBS7110EA 341 EUR
BWRLBS7110
Anti-Ephrin-A1 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human Ephrin-A1.

Ephrin-A1 pAb detects endogenous levels of Ephrin-A1 protein.

Type: Primary
Antigen: Ephrin-A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR