Anti-MgcRacGAP Rabbit Polyclonal Antibody
Lieferant: Bioworld Technology
BS7163
BWRLBS7163EA 341 EUR
BWRLBS7163
Anti-MgcRacGAP Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human MgcRacGAP.
MgcRacGAP pAb detects endogenous levels of MgcRacGAP protein.
Type: Primary
Antigen: MgcRacGAP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat