Anti-MMP-11 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7168
BWRLBS7168EA 341 EUR
BWRLBS7168
Anti-MMP-11 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human MMP-11.

MMP-11 pAb detects endogenous levels of MMP-11 protein.

Type: Primary
Antigen: MMP-11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR