Anti-p68 RNA Helicase Rabbit Polyclonal Antibody
Lieferant: Bioworld Technology
BS7186
BWRLBS7186EA 341 EUR
BWRLBS7186
Anti-p68 RNA Helicase Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human p68 RNA Helicase.
p68 RNA Helicase pAb detects endogenous levels of p68 RNA Helicase protein.
Type: Primary
Antigen: p68 RNA Helicase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat