Anti-Peroxin 19 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7195
BWRLBS7195EA 341 EUR
BWRLBS7195
Anti-Peroxin 19 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human Peroxin 19.

Peroxin 19 pAb detects endogenous levels of Peroxin 19 protein.

Type: Primary
Antigen: Peroxin 19
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR