Anti-RPL5 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS7217
BWRLBS7217EA 341 EUR
BWRLBS7217
Anti-RPL5 Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human RPL5.

RPL5 pAb detects endogenous levels of RPL5 protein.

Type: Primary
Antigen: RPL5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR