Anti-DNMT3L Rabbit Polyclonal Antibody [clone: ChIP Grade]

Lieferant: Bioworld Technology
BS7226
BWRLBS7226EA 341 EUR
BWRLBS7226
Anti-DNMT3L Rabbit Polyclonal Antibody [clone: ChIP Grade]
Antikörper
Recombinant full length Human DNMT3L.

DNMT3L pAb detects endogenous levels of DNMT3L protein.

Type: Primary
Antigen: DNMT3L
Clonality: Polyclonal
Clone: ChIP Grade
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR