Anti-UHRF1 Rabbit Polyclonal Antibody [clone: ChIP Grade]

Lieferant: Bioworld Technology
BS7227
BWRLBS7227EA 341 EUR
BWRLBS7227
Anti-UHRF1 Rabbit Polyclonal Antibody [clone: ChIP Grade]
Antikörper
Recombinant full length Human UHRF1.

UHRF1 pAb detects endogenous levels of UHRF1 protein.

Type: Primary
Antigen: UHRF1
Clonality: Polyclonal
Clone: ChIP Grade
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR